siRNA Details
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033963.1 | NP_001029135.1 | 2 | 447 | ||
GenBank mRNA | BC091203.1 | - | 2 | 447 | 7380393 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033963.1 | NP_001029135.1 | 2 | 447 | ||
GenBank mRNA | BC091203.1 | - | 2 | 447 | 7380393 |