siRNA Details
| Gene Symbol: Prkx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033963.1 | NP_001029135.1 | 1 | 400 | ||
| GenBank mRNA | BC091203.1 | - | 1 | 400 | 7380393 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkx| Gene Symbol: Prkx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033963.1 | NP_001029135.1 | 1 | 400 | ||
| GenBank mRNA | BC091203.1 | - | 1 | 400 | 7380393 | |