siRNA Details
| Gene Symbol: Rfk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014106.1 | NP_001014128.1 | 4 | 1175 | ||
| GenBank mRNA | BC079125.1 | - | 4 | 1175 | 7127049 | |
| FQ219206.1 | - | 4 | 1143 | |||
| FQ223869.1 | - | 4 | 1188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rfk| Gene Symbol: Rfk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014106.1 | NP_001014128.1 | 4 | 1175 | ||
| GenBank mRNA | BC079125.1 | - | 4 | 1175 | 7127049 | |
| FQ219206.1 | - | 4 | 1143 | |||
| FQ223869.1 | - | 4 | 1188 | |||