siRNA Details
| Gene Symbol: RGD1564955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257545.1 | XP_006257607.1 | 3 | 318 | ||
| XM_006257547.2 | XP_006257609.1 | 3 | 317 | |||
| XM_008773543.1 | XP_008771765.1 | 3 | 317 | |||
| XM_008773544.1 | XP_008771766.1 | 3 | 317 | |||
| XM_008773545.1 | XP_008771767.1 | 3 | 361 | |||
| GenBank mRNA | BC160914.1 | - | 3 | 324 | ||

RGD1564955
Genomic Map