siRNA Details
| Gene Symbol: Ctag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109322.1 | NP_001102792.1 | 1 | 194 | ||
| XM_008773587.1 | XP_008771809.1 | 1 | 176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctag2| Gene Symbol: Ctag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109322.1 | NP_001102792.1 | 1 | 194 | ||
| XM_008773587.1 | XP_008771809.1 | 1 | 176 | |||