siRNA Details
| Gene Symbol: Slc10a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024368.1 | NP_001019539.1 | 3 | 978 | ||
| XM_006229606.2 | XP_006229668.1 | 3 | 1735 | |||
| XM_008773611.1 | XP_008771833.1 | 3 | 1118 | |||
| GenBank mRNA | BC086556.1 | - | 3 | 978 | 7304843 | |

Slc10a3
Genomic Map