siRNA Details
| Gene Symbol: Mis12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047972.1 | NP_001041437.1 | 2 | 116 | ||
| XM_006246791.2 | XP_006246853.1 | 3 | 508 | |||
| XM_006246793.2 | XP_006246855.1 | 2 | 224 | |||
| XM_006246794.2 | XP_006246856.1 | 2 | 347 | |||
| GenBank mRNA | AY310159.2 | - | 2 | 116 | ||

Mis12
Genomic Map