siRNA Details
| Gene Symbol: LOC100363287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727853.3 | XP_002727899.3 | 1 | 501 | ||
| Gene Symbol: Krtap13-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109325.1 | NP_001102795.1 | 1 | 537 | ||

LOC100363287
Genomic Map