siRNA Details
| Gene Symbol: NCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199509.1 | NP_001186438.1 | 2 | 431 | ||
| NM_001199510.1 | NP_001186439.1 | 2 | 431 | |||
| NM_004828.3 | NP_004819.2 | 2 | 431 | |||
| XM_017011500.1 | XP_016866989.1 | 2 | 601 | |||
| GenBank mRNA | AJ010099.1 | - | 2 | 370 | ||
| AJ010100.1 | - | 2 | 370 | |||

NCR2
Genomic Map