siRNA Details
| Gene Symbol: Prr36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248788.2 | XP_006248850.1 | 5 | 2293 | ||
| XM_008769018.1 | XP_008767240.1 | 5 | 2635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr36| Gene Symbol: Prr36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248788.2 | XP_006248850.1 | 5 | 2293 | ||
| XM_008769018.1 | XP_008767240.1 | 5 | 2635 | |||