siRNA Details
| Gene Symbol: LOC102554944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248802.2 | XP_006248864.1 | 2 | 200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102554944| Gene Symbol: LOC102554944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248802.2 | XP_006248864.1 | 2 | 200 | ||