siRNA Details
| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192005.1 | NP_001178934.1 | 6 | 563 | ||
| XM_006248775.2 | XP_006248837.1 | 5 | 1176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd209e| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192005.1 | NP_001178934.1 | 6 | 563 | ||
| XM_006248775.2 | XP_006248837.1 | 5 | 1176 | |||