siRNA Details
Gene Symbol: Cd209e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192005.1 | NP_001178934.1 | 6 | 539 | ||
XM_006248775.2 | XP_006248837.1 | 5 | 1152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd209e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192005.1 | NP_001178934.1 | 6 | 539 | ||
XM_006248775.2 | XP_006248837.1 | 5 | 1152 |