siRNA Details
| Gene Symbol: Ksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249485.2 | XP_006249547.1 | 1 | 784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ksr2| Gene Symbol: Ksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249485.2 | XP_006249547.1 | 1 | 784 | ||