siRNA Details
| Gene Symbol: Rab7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109328.2 | NP_001102798.1 | 2 | 130 | ||
| XM_008769490.1 | XP_008767712.1 | 2 | 199 | |||
| XM_008769491.1 | XP_008767713.1 | 2 | 545 | |||
| XM_008769492.1 | XP_008767714.1 | 2 | 546 | |||
| XM_008769493.1 | XP_008767715.1 | 2 | 550 | |||
| GenBank mRNA | FM076322.1 | - | 2 | 130 | ||

Rab7b
Genomic Map