siRNA Details
| Gene Symbol: Rab7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109328.2 | NP_001102798.1 | 4 | 482 | ||
| XM_008769490.1 | XP_008767712.1 | 4 | 551 | |||
| XM_008769491.1 | XP_008767713.1 | 4 | 897 | |||
| XM_008769492.1 | XP_008767714.1 | 4 | 898 | |||
| XM_008769493.1 | XP_008767715.1 | 4 | 902 | |||
| GenBank mRNA | FM076322.1 | - | 4 | 482 | ||

Rab7b
Genomic Map