siRNA Details
Gene Symbol: LOC100910879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250214.2 | XP_006250276.1 | 1 | 237 |
Gene Symbol: Sh2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_577285.3 | XP_577285.1 | 2 | 1857 |
