siRNA Details
| Gene Symbol: RGD1561699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250714.2 | XP_006250776.1 | 2 | 755 | ||
| XM_008770085.1 | XP_008768307.1 | 2 | 752 | |||
| Gene Symbol: LOC100910292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250720.1 | XP_006250782.1 | 2 | 752 | ||
| Gene Symbol: LOC100912233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250508.1 | XP_006250570.1 | 2 | 755 | ||
| Gene Symbol: RGD1562423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250509.1 | XP_006250571.1 | 755 | |||

RGD1561699
Genomic Map