siRNA Details
Gene Symbol: RGD1565660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751332.1 | XP_003751380.1 | 1 | 258 | ||
XM_003751333.1 | XP_003751381.1 | 1 | 258 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1565660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751332.1 | XP_003751380.1 | 1 | 258 | ||
XM_003751333.1 | XP_003751381.1 | 1 | 258 |