siRNA Details
| Gene Symbol: RGD1565660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751332.1 | XP_003751380.1 | 3 | 995 | ||
| XM_003751333.1 | XP_003751381.1 | 2 | 407 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565660| Gene Symbol: RGD1565660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751332.1 | XP_003751380.1 | 3 | 995 | ||
| XM_003751333.1 | XP_003751381.1 | 2 | 407 | |||