siRNA Details
| Gene Symbol: RGD1561226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109329.1 | NP_001102799.1 | 3 | 1037 | ||
| XM_006250716.1 | XP_006250778.1 | 2 | 443 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561226| Gene Symbol: RGD1561226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109329.1 | NP_001102799.1 | 3 | 1037 | ||
| XM_006250716.1 | XP_006250778.1 | 2 | 443 | |||