siRNA Details
| Gene Symbol: LOC100909497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770129.1 | XP_008768351.1 | 4 | 1205 | ||
| Gene Symbol: LOC685614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769913.1 | XP_008768135.1 | 3 | 1355 | ||

LOC100909497
Genomic Map