siRNA Details
| Gene Symbol: Cbln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109330.1 | NP_001102800.1 | 1 | 440 | ||
| XM_006252007.2 | XP_006252069.1 | 1 | 530 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbln3| Gene Symbol: Cbln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109330.1 | NP_001102800.1 | 1 | 440 | ||
| XM_006252007.2 | XP_006252069.1 | 1 | 530 | |||