siRNA Details
| Gene Symbol: Gzmbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773505.1 | XP_008771727.1 | 2 | 223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gzmbl1| Gene Symbol: Gzmbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773505.1 | XP_008771727.1 | 2 | 223 | ||