siRNA Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134727.1 | NP_001128199.1 | 11, 12 | 1526 | ||
| XM_006252311.2 | XP_006252373.1 | 11, 12 | 1608 | |||
| XM_006252312.2 | XP_006252374.1 | 11, 12 | 1602 | |||
| XM_006252313.2 | XP_006252375.1 | 11, 12 | 1604 | |||
| XM_006252315.2 | XP_006252377.1 | 11, 12 | 1604 | |||
| GenBank mRNA | BC166585.1 | - | 11, 12 | 1526 | 7444640 | |

Lrch1
Genomic Map