siRNA Details
| Gene Symbol: RGD1562508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771207.1 | XP_008769429.1 | 2 | 838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562508| Gene Symbol: RGD1562508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771207.1 | XP_008769429.1 | 2 | 838 | ||