siRNA Details
| Gene Symbol: RGD1562638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100944.1 | NP_001094414.1 | 4 | 2868 | ||
| XM_008771316.1 | XP_008769538.1 | 8 | 3674 | |||
| XM_008771317.1 | XP_008769539.1 | 7 | 3622 | |||
| XM_008771318.1 | XP_008769540.1 | 9 | 3728 | |||
| XM_008771319.1 | XP_008769541.1 | 8 | 3674 | |||
| XM_008771320.1 | XP_008769542.1 | 6 | 3888 | |||
| XM_008771321.1 | XP_008769543.1 | 5 | 3301 | |||
| XM_008771322.1 | XP_008769544.1 | 8 | 3674 | |||
| GenBank mRNA | BC128761.1 | - | 4 | 2868 | 7113007 | |

RGD1562638
Genomic Map