siRNA Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728509.4 | XP_002728555.2 | 2 | 816 | ||
| XM_006254225.2 | XP_006254287.1 | 2 | 811 | |||
| GenBank mRNA | DQ157696.1 | - | 2 | 103 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il15ra| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728509.4 | XP_002728555.2 | 2 | 816 | ||
| XM_006254225.2 | XP_006254287.1 | 2 | 811 | |||
| GenBank mRNA | DQ157696.1 | - | 2 | 103 | ||