siRNA Details
| Gene Symbol: Lvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577617.5 | XP_577617.3 | 4 | 2723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lvrn| Gene Symbol: Lvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577617.5 | XP_577617.3 | 4 | 2723 | ||