siRNA Details
| Gene Symbol: Lrrc29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195616.1 | NP_001182545.1 | 3 | 1067 | ||
| XM_006255531.2 | XP_006255593.1 | 3 | 1302 | |||
| XM_006255532.2 | XP_006255594.1 | 3 | 1210 | |||
| XM_006255533.2 | XP_006255595.1 | 3 | 1146 | |||
| GenBank mRNA | BC166813.1 | - | 3 | 1067 | 7454853 | |

Lrrc29
Genomic Map