siRNA Details
| Gene Symbol: LOC100361016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773378.1 | XP_008771600.1 | 1 | 1511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100361016| Gene Symbol: LOC100361016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773378.1 | XP_008771600.1 | 1 | 1511 | ||