siRNA Details
| Gene Symbol: GSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191002.1 | NP_001177931.1 | 5 | 804 | ||
| NM_001191003.1 | NP_001177932.1 | 6 | 956 | |||
| NM_004832.2 | NP_004823.1 | 6 | 903 | |||
| GenBank mRNA | AF212303.1 | - | 6 | 710 | ||
| AK313680.1 | - | 6 | 721 | |||
| BC000127.1 | - | 6 | 713 | 3351359 | ||
| BX537431.1 | - | 6 | 831 | |||
| CF130513.1 | - | 6 | 679 | |||
| CN283947.1 | - | 5 | 607 | |||
| EU794619.1 | - | 6 | 710 | |||
| U90313.1 | - | 6 | 693 | |||

GSTO1
Genomic Map