siRNA Details
| Gene Symbol: AIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004833.1 | NP_004824.1 | 5 | 1172 | ||
| XM_005245616.4 | XP_005245673.1 | 4 | 943 | |||
| XM_017002848.1 | XP_016858337.1 | 6 | 2399 | |||
| GenBank mRNA | AF024714.1 | - | 5 | 1172 | ||
| AK292042.1 | - | 5 | 1081 | |||
| BC010940.1 | - | 5 | 1174 | 4080360 | ||

AIM2
Genomic Map