siRNA Details
| Gene Symbol: Stard5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192011.1 | NP_001178940.1 | 4 | 415 | ||
| GenBank mRNA | FQ230578.1 | - | 4 | 389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stard5| Gene Symbol: Stard5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192011.1 | NP_001178940.1 | 4 | 415 | ||
| GenBank mRNA | FQ230578.1 | - | 4 | 389 | ||