siRNA Details
| Gene Symbol: RGD1562885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166267.1 | NP_001159739.1 | 2 | 233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562885| Gene Symbol: RGD1562885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166267.1 | NP_001159739.1 | 2 | 233 | ||