siRNA Details
| Gene Symbol: RGD1559903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109337.1 | NP_001102807.1 | 1 | 214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559903| Gene Symbol: RGD1559903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109337.1 | NP_001102807.1 | 1 | 214 | ||