siRNA Details
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037656.1 | NP_001032745.1 | 8 | 1268 | ||
GenBank mRNA | BC107937.1 | - | 8 | 1268 | 7320504 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037656.1 | NP_001032745.1 | 8 | 1268 | ||
GenBank mRNA | BC107937.1 | - | 8 | 1268 | 7320504 |