siRNA Details
| Gene Symbol: RGD1561998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578019.5 | XP_578019.5 | 3 | 375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561998| Gene Symbol: RGD1561998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578019.5 | XP_578019.5 | 3 | 375 | ||