siRNA Details
| Gene Symbol: LOC689173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750755.3 | XP_003750803.1 | 1602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC689173| Gene Symbol: LOC689173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750755.3 | XP_003750803.1 | 1602 | |||