siRNA Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035255.1 | NP_001030332.1 | 2 | 401 | ||
| XM_006233540.2 | XP_006233602.1 | 3 | 441 | |||
| XM_006233541.2 | XP_006233603.1 | 3 | 440 | |||
| XM_006233542.2 | XP_006233604.1 | 3 | 441 | |||
| XM_006233543.2 | XP_006233605.1 | 4 | 498 | |||
| XM_008761567.1 | XP_008759789.1 | 3 | 442 | |||
| XM_008761568.1 | XP_008759790.1 | 4 | 3489 | |||
| XM_008761569.1 | XP_008759791.1 | 4 | 505 | |||
| GenBank mRNA | BC099157.1 | - | 2 | 401 | 7381485 | |
| FQ209707.1 | - | 3 | 407 | |||
| FQ234514.1 | - | 2 | 416 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001093752, NM_001093753, BC069945, AK040467, AK011472, and others.

Srsf11
Genomic Map