siRNA Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035255.1 | NP_001030332.1 | 8 | 966 | ||
| XM_006233540.2 | XP_006233602.1 | 9 | 1006 | |||
| XM_006233541.2 | XP_006233603.1 | 9 | 1005 | |||
| XM_006233542.2 | XP_006233604.1 | 9 | 1006 | |||
| XM_006233543.2 | XP_006233605.1 | 10 | 1063 | |||
| XM_008761567.1 | XP_008759789.1 | 9 | 1007 | |||
| XM_008761568.1 | XP_008759790.1 | 10 | 4054 | |||
| XM_008761569.1 | XP_008759791.1 | 10 | 1070 | |||
| XM_008761570.1 | XP_008759792.1 | 6 | 1254 | |||
| GenBank mRNA | BC099157.1 | - | 8 | 966 | 7381485 | |
| FQ234514.1 | - | 8 | 981 | |||

Srsf11
Genomic Map