siRNA Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035255.1 | NP_001030332.1 | 8 | 935 | ||
| XM_006233540.2 | XP_006233602.1 | 9 | 975 | |||
| XM_006233541.2 | XP_006233603.1 | 9 | 974 | |||
| XM_006233542.2 | XP_006233604.1 | 9 | 975 | |||
| XM_006233543.2 | XP_006233605.1 | 10 | 1032 | |||
| XM_008761567.1 | XP_008759789.1 | 9 | 976 | |||
| XM_008761568.1 | XP_008759790.1 | 10 | 4023 | |||
| XM_008761569.1 | XP_008759791.1 | 10 | 1039 | |||
| XM_008761570.1 | XP_008759792.1 | 6 | 1223 | |||
| GenBank mRNA | BC099157.1 | - | 8 | 935 | 7381485 | |
| FQ209707.1 | - | 9 | 941 | |||
| FQ234514.1 | - | 8 | 950 | |||

Srsf11
Genomic Map