siRNA Details
| Gene Symbol: LOC502618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037366.1 | NP_001032443.1 | 5 | 568 | ||
| XM_006234041.2 | XP_006234103.1 | 7 | 773 | |||
| XM_008761772.1 | XP_008759994.1 | 3 | 521 | |||
| GenBank mRNA | AY524049.1 | - | 5 | 568 | ||

LOC502618
Genomic Map