siRNA Details
| Gene Symbol: LOC502618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037366.1 | NP_001032443.1 | 5 | 524 | ||
| GenBank mRNA | AY524049.1 | - | 5 | 524 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC502618| Gene Symbol: LOC502618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037366.1 | NP_001032443.1 | 5 | 524 | ||
| GenBank mRNA | AY524049.1 | - | 5 | 524 | ||