siRNA Details
| Gene Symbol: LOC502660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578154.5 | XP_578154.3 | 1 | 521 | ||
| Gene Symbol: Olr792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000378.1 | NP_001000378.1 | 1 | 488 | ||

LOC502660
Genomic Map