siRNA Details
| Gene Symbol: LOC502660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578154.5 | XP_578154.3 | 1 | 856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC502660| Gene Symbol: LOC502660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578154.5 | XP_578154.3 | 1 | 856 | ||