siRNA Details
| Gene Symbol: LOC502684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025060.1 | NP_001020231.1 | 8 | 939 | ||
| XM_006235246.2 | XP_006235308.1 | 8 | 980 | |||
| XM_006235247.2 | XP_006235309.1 | 8 | 978 | |||
| XM_006235248.2 | XP_006235310.1 | 8 | 1079 | |||
| XM_006235249.2 | XP_006235311.1 | 9 | 957 | |||
| XM_006235251.2 | XP_006235313.1 | 8 | 979 | |||
| XM_008762376.1 | XP_008760598.1 | 8 | 1058 | |||
| XM_008762377.1 | XP_008760599.1 | 9 | 1017 | |||
| XM_008762378.1 | XP_008760600.1 | 8 | 959 | |||
| GenBank mRNA | BC083680.1 | - | 8 | 939 | 7135883 | |

LOC502684
Genomic Map