siRNA Details
Gene Symbol: Tldc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235394.2 | XP_006235456.1 | 1 | 355 | ||
XM_578186.6 | XP_578186.4 | 1 | 355 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tldc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235394.2 | XP_006235456.1 | 1 | 355 | ||
XM_578186.6 | XP_578186.4 | 1 | 355 |