siRNA Details
| Gene Symbol: RGD1561560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235972.2 | XP_006236034.1 | 2 | 661 | ||
| XM_578210.6 | XP_578210.4 | 2 | 516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561560| Gene Symbol: RGD1561560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235972.2 | XP_006236034.1 | 2 | 661 | ||
| XM_578210.6 | XP_578210.4 | 2 | 516 | |||