siRNA Details
| Gene Symbol: Lrrc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025155.2 | NP_001020326.1 | 4 | 1505 | ||
| XM_008762657.1 | XP_008760879.1 | 4 | 1793 | |||
| GenBank mRNA | BC079098.1 | - | 4 | 1382 | 7124390 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc17| Gene Symbol: Lrrc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025155.2 | NP_001020326.1 | 4 | 1505 | ||
| XM_008762657.1 | XP_008760879.1 | 4 | 1793 | |||
| GenBank mRNA | BC079098.1 | - | 4 | 1382 | 7124390 | |